Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609870 | ||||||||||||||||||||
Literature Links: |
ARHGAP21 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGAP21 - Rho GTPase activating protein 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020824.3 | 7338 | Intron | NP_065875.3 | |||
XM_005252542.3 | 7338 | Intron | XP_005252599.1 | |||
XM_005252544.4 | 7338 | UTR 3 | XP_005252601.1 | |||
XM_011519602.2 | 7338 | UTR 3 | XP_011517904.1 | |||
XM_011519603.2 | 7338 | UTR 3 | XP_011517905.1 | |||
XM_011519604.2 | 7338 | Intron | XP_011517906.1 | |||
XM_011519605.2 | 7338 | UTR 3 | XP_011517907.1 | |||
XM_011519606.2 | 7338 | UTR 3 | XP_011517908.1 | |||
XM_011519607.2 | 7338 | UTR 3 | XP_011517909.1 | |||
XM_017016459.1 | 7338 | UTR 3 | XP_016871948.1 | |||
XM_017016460.1 | 7338 | UTR 3 | XP_016871949.1 | |||
XM_017016461.1 | 7338 | Intron | XP_016871950.1 |