Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAACAATTTTAACATGGTTTATTA[C/T]GTTTCCAGAAAGGTTAAGTCTTATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
39 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927560 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC101927560 - uncharacterized LOC101927560 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101927587 - uncharacterized LOC101927587 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTLL7 - tubulin tyrosine ligase like 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024686.4 | 3068 | UTR 3 | NP_078962.4 | |||
XM_005271208.2 | 3068 | UTR 3 | XP_005271265.1 | |||
XM_011542159.2 | 3068 | Intron | XP_011540461.1 | |||
XM_017002347.1 | 3068 | UTR 3 | XP_016857836.1 | |||
XM_017002348.1 | 3068 | Intron | XP_016857837.1 | |||
XM_017002349.1 | 3068 | UTR 3 | XP_016857838.1 |