Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTCCAGCCGGGGCCCTGTCTTC[T/C]AGATCAGGGTTGGCTGCTGTCTATC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 190170 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TGFA PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TGFA - transforming growth factor alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099691.2 | 3577 | UTR 3 | NP_001093161.1 | |||
NM_001308158.1 | 3577 | UTR 3 | NP_001295087.1 | |||
NM_001308159.1 | 3577 | UTR 3 | NP_001295088.1 | |||
NM_003236.3 | 3577 | UTR 3 | NP_003227.1 |
Set Membership: |
HapMap |