Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCCCATCCCTGGCTTGGTTCCA[C/T]GTGTGTCCAGCCGGGGCCCTGTCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 190170 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TGFA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TGFA - transforming growth factor alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099691.2 | 3604 | UTR 3 | NP_001093161.1 | |||
NM_001308158.1 | 3604 | UTR 3 | NP_001295087.1 | |||
NM_001308159.1 | 3604 | UTR 3 | NP_001295088.1 | |||
NM_003236.3 | 3604 | UTR 3 | NP_003227.1 |