Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAGGGCTTCAGCCGCAGCACGGAC[C/T]TGGTGCGCCACCAGGCCACGCACAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF853 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF853 - zinc finger protein 853 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017560.1 | 1939 | Silent Mutation | CTG,TTG | L,L 500 | NP_060030.1 | |
XM_011515438.2 | 1939 | Silent Mutation | CTG,TTG | L,L 526 | XP_011513740.1 | |
XM_011515439.2 | 1939 | Silent Mutation | CTG,TTG | L,L 511 | XP_011513741.1 | |
XM_017012371.1 | 1939 | Silent Mutation | CTG,TTG | L,L 485 | XP_016867860.1 |