Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGAGGTGAAAATTCCCCAGAATT[A/G]TTGACACACACGGCGGCACACCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118493 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHRM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHRM2 - cholinergic receptor muscarinic 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000739.2 | Intron | NP_000730.1 | ||||
NM_001006626.1 | Intron | NP_001006627.1 | ||||
NM_001006627.1 | Intron | NP_001006628.1 | ||||
NM_001006628.1 | Intron | NP_001006629.1 | ||||
NM_001006629.1 | Intron | NP_001006630.1 | ||||
NM_001006630.1 | Intron | NP_001006631.1 | ||||
NM_001006631.1 | Intron | NP_001006632.1 | ||||
NM_001006632.1 | Intron | NP_001006633.1 | ||||
XM_011515769.2 | Intron | XP_011514071.1 |