Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCCACAACTGAACACCACCAACG[G/A]AAATGGCCACGCCGGAACCGGAACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 123996 MIM: 611028 | ||||||||||||||||||||
Literature Links: |
COX7A2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COX7A2 - cytochrome c oxidase subunit 7A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001865.3 | 147 | Missense Mutation | CCG,TCG | P,S 17 | NP_001856.2 |
TMEM30A - transmembrane protein 30A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |