Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGCAAAGCAGTGGGGATTGTGCC[C/T]GGGCCAAACACACAGGAACCAGGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613421 MIM: 614636 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCTD10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCTD10 - potassium channel tetramerization domain containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317395.1 | 1741 | UTR 3 | NP_001304324.1 | |||
NM_001317399.1 | 1741 | UTR 3 | NP_001304328.1 | |||
NM_031954.4 | 1741 | UTR 3 | NP_114160.1 |
MYO1H - myosin IH | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |