Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACCGCGCGGGACGGAGCTTGGCT[G/T]TTGGTCGGTGGGTTCCCGTGCGGCG
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 602688 MIM: 614683 | ||||||||||||||||||||||||||
Literature Links: |
HNRNPAB PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
HNRNPAB - heterogeneous nuclear ribonucleoprotein A/B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004499.3 | 60 | UTR 5 | NP_004490.2 | |||
NM_031266.2 | 60 | UTR 5 | NP_112556.2 |
PHYKPL - 5-phosphohydroxy-L-lysine phospho-lyase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |