Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTCCTCCTCATCCTCTGAGGCA[G/A]CCATCGCTCCTGGAGGTTCTGGCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610052 | ||||||||||||||||||||
Literature Links: |
CHMP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHMP3 - charged multivesicular body protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005753.2 | 681 | Missense Mutation | GCT,GTT | A,V 132 | NP_001005753.1 | |
NM_001193517.1 | 681 | Missense Mutation | GCT,GTT | A,V 158 | NP_001180446.1 | |
NM_016079.3 | 681 | Missense Mutation | GCT,GTT | A,V 198 | NP_057163.1 |
RNF103-CHMP3 - RNF103-CHMP3 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198954.1 | 681 | Missense Mutation | GCT,GTT | A,V 227 | NP_001185883.1 |