Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCATTTGGGCACATGGTGTGCCC[C/T]CTCCACTAGCCTGGCATCTCAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608302 MIM: 609349 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LGI3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LGI3 - leucine rich repeat LGI family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139278.2 | 3156 | UTR 3 | NP_644807.1 |
REEP4 - receptor accessory protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |