Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCACCCCCAGAACTAGAGCTTTCC[C/T]AGCTGTAAGCCTTATAAACCAAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
22 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612257 MIM: 606076 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAST2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAST2 - microtubule associated serine/threonine kinase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIK3R3 - phosphoinositide-3-kinase regulatory subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114172.1 | 3164 | UTR 3 | NP_001107644.1 | |||
NM_001303427.1 | 3164 | UTR 3 | NP_001290356.1 | |||
NM_001303428.1 | 3164 | UTR 3 | NP_001290357.1 | |||
NM_001303429.1 | 3164 | UTR 3 | NP_001290358.1 | |||
NM_003629.3 | 3164 | UTR 3 | NP_003620.3 | |||
XM_017002614.1 | 3164 | UTR 3 | XP_016858103.1 |