Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTACCCAAAAGTTTTTGAGTAAAC[C/T]GAAAGTTAGATACTCCAGGAACAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603561 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MTMR6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MTMR6 - myotubularin related protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004685.3 | 2448 | UTR 3 | NP_004676.3 | |||
XM_011535306.1 | 2448 | UTR 3 | XP_011533608.1 | |||
XM_011535307.1 | 2448 | UTR 3 | XP_011533609.1 | |||
XM_017020845.1 | 2448 | UTR 3 | XP_016876334.1 | |||
XM_017020846.1 | 2448 | UTR 3 | XP_016876335.1 |