Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGTGAGGATTCCACGGTGTTGGG[T/G]AGGCAGGCAGGGTGGTACTCATGAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605564 | ||||||||||||||||||||
Literature Links: |
CIB2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CIB2 - calcium and integrin binding family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271888.1 | 1294 | UTR 3 | NP_001258817.1 | |||
NM_001271889.1 | 1294 | UTR 3 | NP_001258818.1 | |||
NM_001301224.1 | 1294 | UTR 3 | NP_001288153.1 | |||
NM_006383.3 | 1294 | UTR 3 | NP_006374.1 | |||
XM_005254126.3 | 1294 | Intron | XP_005254183.1 | |||
XM_006720374.2 | 1294 | UTR 3 | XP_006720437.1 | |||
XM_011521161.2 | 1294 | Intron | XP_011519463.1 |
SH2D7 - SH2 domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101404.1 | 1294 | Intron | NP_001094874.1 | |||
XM_011521895.2 | 1294 | Intron | XP_011520197.1 | |||
XM_011521896.2 | 1294 | Intron | XP_011520198.1 | |||
XM_011521899.2 | 1294 | Intron | XP_011520201.1 | |||
XM_017022487.1 | 1294 | Intron | XP_016877976.1 | |||
XM_017022488.1 | 1294 | Intron | XP_016877977.1 |