Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCCTCTGGGGAGCGACCGCAGGC[T/C]AACCAGAAGCGAATCACCACACCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604414 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C22orf23 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C22orf23 - chromosome 22 open reading frame 23 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6820 - microRNA 6820 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLR2F - RNA polymerase II subunit F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301129.1 | 266 | Silent Mutation | GCC,GCT | A,A 38 | NP_001288058.1 | |
NM_001301130.1 | 266 | Silent Mutation | GCC,GCT | A,A 47 | NP_001288059.1 | |
NM_001301131.1 | 266 | Silent Mutation | GCC,GCT | A,A 47 | NP_001288060.1 | |
NM_021974.4 | 266 | Silent Mutation | GCC,GCT | A,A 47 | NP_068809.1 | |
XM_017028832.1 | 266 | Silent Mutation | GCC,GCT | A,A 47 | XP_016884321.1 |