Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAATGTGTCCCTGTTAACTGACT[A/G]TATTTATTAAATTGAATTAAGGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300160 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDX3X PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDX3X - DEAD-box helicase 3, X-linked | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193416.2 | 1305 | Intron | NP_001180345.1 | |||
NM_001193417.2 | 1305 | Intron | NP_001180346.1 | |||
NM_001356.4 | 1305 | Intron | NP_001347.3 | |||
XM_011543892.2 | 1305 | Intron | XP_011542194.1 | |||
XM_017029313.1 | 1305 | UTR 5 | XP_016884802.1 | |||
XM_017029314.1 | 1305 | UTR 5 | XP_016884803.1 |
LOC107985678 - uncharacterized LOC107985678 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017030007.1 | 1305 | Intron | XP_016885496.1 |