Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCTTCTTACTATGCGAACAGAGG[A/C]AGAGGAGGGATAGAGGCCCAGCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609359 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HS1BP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HS1BP3 - HCLS1 binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022460.3 | 1648 | UTR 3 | NP_071905.3 | |||
XM_017004696.1 | 1648 | Intron | XP_016860185.1 | |||
XM_017004697.1 | 1648 | Intron | XP_016860186.1 | |||
XM_017004698.1 | 1648 | Intron | XP_016860187.1 | |||
XM_017004699.1 | 1648 | Intron | XP_016860188.1 | |||
XM_017004700.1 | 1648 | UTR 3 | XP_016860189.1 | |||
XM_017004701.1 | 1648 | Intron | XP_016860190.1 |