Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTGCCGTCGCCACTCTTGCTGTC[G/C]GTGATGTGCAGGGAAAGCACACCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605193 MIM: 615406 | ||||||||||||||||||||
Literature Links: |
DIRAS3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DIRAS3 - DIRAS family GTPase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004675.3 | 1550 | Silent Mutation | ACC,ACG | T,T 90 | NP_004666.1 | |
XM_017002773.1 | 1550 | Silent Mutation | ACC,ACG | T,T 90 | XP_016858262.1 |
GNG12-AS1 - GNG12 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |