Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAGGCTGTCGGGCGTCATGGTGC[C/G]GGCGCCCATCCAAGACCTGGAGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611150 | ||||||||||||||||||||
Literature Links: |
ATXN10 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATXN10 - ataxin 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167621.1 | 316 | Missense Mutation | CCG,CGG | P,R 17 | NP_001161093.1 | |
NM_013236.3 | 316 | Missense Mutation | CCG,CGG | P,R 17 | NP_037368.1 |