Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAGGGACTTTTGATGTATTTGAA[C/T]CCACCTCCTTCTGAAAGCAGGAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138300 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GSR - glutathione-disulfide reductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000637.3 | 2767 | UTR 3 | NP_000628.2 | |||
NM_001195102.1 | 2767 | UTR 3 | NP_001182031.1 | |||
NM_001195103.1 | 2767 | UTR 3 | NP_001182032.1 | |||
NM_001195104.1 | 2767 | UTR 3 | NP_001182033.1 |