Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTGTGCCTGGCCTTCCAAGGCA[C/T]ACCTGTCACTGGCTTGGAACTCAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602349 MIM: 613577 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NTN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NTN3 - netrin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBC1D24 - TBC1 domain family member 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199107.1 | Intron | NP_001186036.1 | ||||
NM_020705.2 | Intron | NP_065756.1 | ||||
XM_017023493.1 | Intron | XP_016878982.1 | ||||
XM_017023494.1 | Intron | XP_016878983.1 | ||||
XM_017023495.1 | Intron | XP_016878984.1 |