Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCTCCCAGTTCAGGAAGTCCACC[T/C]GCTTCAGCAGCTTCTGCTCGTGGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612980 | ||||||||||||||||||||
Literature Links: |
IMP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IMP3 - IMP3, U3 small nucleolar ribonucleoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018285.3 | 199 | Missense Mutation | CAG,CGG | Q,R 15 | NP_060755.1 |
LOC107984799 - translation initiation factor IF-2-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNX33 - sorting nexin 33 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |