Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGTTCGGAACGGACCCCCGGGGC[C/T]GCAACCGCCAGCGCCTCAGGGGCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600798 MIM: 608061 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NECTIN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NECTIN2 - nectin cell adhesion molecule 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TOMM40 - translocase of outer mitochondrial membrane 40 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128916.1 | Intron | NP_001122388.1 | ||||
NM_001128917.1 | Intron | NP_001122389.1 | ||||
NM_006114.2 | Intron | NP_006105.1 | ||||
XM_005258411.3 | Intron | XP_005258468.1 |