Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTCTTCTTCTAAGGCTTAGGTTT[T/C]GCCCAGAATTCCCGATACATGGAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
HIGD1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HIGD1A - HIG1 hypoxia inducible domain family member 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099668.1 | 389 | Silent Mutation | GCA,GCG | A,A 103 | NP_001093138.1 | |
NM_001099669.1 | 389 | Silent Mutation | GCA,GCG | A,A 89 | NP_001093139.1 | |
NM_014056.3 | 389 | Silent Mutation | GCA,GCG | A,A 89 | NP_054775.2 |