Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCTGGCAGCGGCGTCTGCGCGCGG[A/C]ATAACGACCCAGGTCGCGGCGCGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 136510 | ||||||||||||||||||||
Literature Links: |
FPGS PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FPGS - folylpolyglutamate synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018078.2 | 130 | Intron | NP_001018088.1 | |||
NM_001288803.1 | 130 | Silent Mutation | GGA,GGC | G,G 21 | NP_001275732.1 | |
NM_004957.5 | 130 | Silent Mutation | GGA,GGC | G,G 21 | NP_004948.4 |