Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGATAAGATCAATGCCCTCATTAA[A/G]GCAGCCGGTGTAAATGTTGAGCCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605064 MIM: 180520 | ||||||||||||||||||||
Literature Links: |
KIF23 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIF23 - kinesin family member 23 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPLP1 - ribosomal protein lateral stalk subunit P1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003.2 | Intron | NP_000994.1 | ||||
NM_213725.1 | Intron | NP_998890.1 |