Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGGCTGGGTGGCAGGCCCCTTG[A/G]TCAAGGTCACAAATCAAGGAGCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607988 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPOCK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPOCK2 - sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134434.1 | 4394 | Intron | NP_001127906.1 | |||
NM_001244950.1 | 4394 | UTR 3 | NP_001231879.1 | |||
NM_014767.2 | 4394 | UTR 3 | NP_055582.1 | |||
XM_011540404.2 | 4394 | UTR 3 | XP_011538706.1 | |||
XM_017016985.1 | 4394 | Intron | XP_016872474.1 |