Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604914 MIM: 615262 MIM: 600813 | ||||||||||||||||||||
Literature Links: |
JMJD6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
JMJD6 - arginine demethylase and lysine hydroxylase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
METTL23 - methyltransferase like 23 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR636 - microRNA 636 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SRSF2 - serine and arginine rich splicing factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195427.1 | 690 | Missense Mutation | CCG,TCG | P,S 147 | NP_001182356.1 | |
NM_003016.4 | 690 | Missense Mutation | CCG,TCG | P,S 147 | NP_003007.2 | |
XM_017024942.1 | 690 | Missense Mutation | CCG,TCG | P,S 147 | XP_016880431.1 |