Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCACATCTACATACTATTTTGCTA[G/T]TCTACATGGGTACATTATTTCCAAC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 606347 MIM: 613134 | |||||||||||||||||||||||
Literature Links: |
PSTPIP1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
PSTPIP1 - proline-serine-threonine phosphatase interacting protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TSPAN3 - tetraspanin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168412.1 | 1766 | UTR 3 | NP_001161884.1 | |||
NM_005724.5 | 1766 | UTR 3 | NP_005715.1 | |||
NM_198902.2 | 1766 | UTR 3 | NP_944492.1 | |||
XM_017021857.1 | 1766 | UTR 3 | XP_016877346.1 |