Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCAACGATCGGAACGAAACTTGG[C/T]TGTGCTTCACCGTGGAAGGTATAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607750 MIM: 609900 MIM: 608993 | ||||||||||||||||||||
Literature Links: |
APOBEC3C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APOBEC3C - apolipoprotein B mRNA editing enzyme catalytic subunit 3C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014508.2 | 206 | Silent Mutation | CTG,TTG | L,L 35 | NP_055323.2 |
APOBEC3D - apolipoprotein B mRNA editing enzyme catalytic subunit 3D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
APOBEC3F - apolipoprotein B mRNA editing enzyme catalytic subunit 3F | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |