Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTTGTCTTCACACAGCAGCGGCA[T/G]GTGCAGAATGATCTCCAGTGGCTCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601304 | ||||||||||||||||||||
Literature Links: |
SNU13 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SNU13 - SNU13 homolog, small nuclear ribonucleoprotein (U4/U6.U5) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003796.1 | 318 | Missense Mutation | ATG,CTG | M,L 69 | NP_001003796.1 | |
NM_005008.3 | 318 | Missense Mutation | ATG,CTG | M,L 69 | NP_004999.1 | |
XM_017028811.1 | 318 | Intron | XP_016884300.1 |