Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGATGCCCCAGGGCTGAGGCCTCC[A/G]CTCAGGCAGCCGCATCTGGGGTCAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 131530 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EGF PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
EGF - epidermal growth factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178130.2 | 352 | UTR 5 | NP_001171601.1 | |||
NM_001178131.2 | 352 | UTR 5 | NP_001171602.1 | |||
NM_001963.5 | 352 | UTR 5 | NP_001954.2 | |||
XM_005262796.2 | 352 | UTR 5 | XP_005262853.1 | |||
XM_011531707.1 | 352 | Intron | XP_011530009.1 | |||
XM_017007845.1 | 352 | UTR 5 | XP_016863334.1 | |||
XM_017007846.1 | 352 | UTR 5 | XP_016863335.1 | |||
XM_017007847.1 | 352 | UTR 5 | XP_016863336.1 | |||
XM_017007848.1 | 352 | UTR 5 | XP_016863337.1 | |||
XM_017007849.1 | 352 | UTR 5 | XP_016863338.1 | |||
XM_017007850.1 | 352 | UTR 5 | XP_016863339.1 | |||
XM_017007851.1 | 352 | UTR 5 | XP_016863340.1 | |||
XM_017007852.1 | 352 | UTR 5 | XP_016863341.1 | |||
XM_017007853.1 | 352 | UTR 5 | XP_016863342.1 | |||
XM_017007854.1 | 352 | UTR 5 | XP_016863343.1 | |||
XM_017007855.1 | 352 | UTR 5 | XP_016863344.1 |
Set Membership: |
HapMap |