Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTAACCATCCCCCTTGCGTTTAC[A/G]ATGTTACAAAATTCAACAAAATAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107930 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDC - dopa decarboxylase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000790.3 | Intron | NP_000781.1 | ||||
NM_001082971.1 | Intron | NP_001076440.1 | ||||
NM_001242886.1 | Intron | NP_001229815.1 | ||||
NM_001242887.1 | Intron | NP_001229816.1 | ||||
NM_001242888.1 | Intron | NP_001229817.1 | ||||
NM_001242889.1 | Intron | NP_001229818.1 | ||||
NM_001242890.1 | Intron | NP_001229819.1 | ||||
XM_005271745.4 | Intron | XP_005271802.1 | ||||
XM_011515161.2 | Intron | XP_011513463.2 |