Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGTGTACAGCGCCAGGAGGTCCA[G/T]CAGCAGCAGGCGGCGCTGGCGCGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 615000 | ||||||||||||||||||||
Literature Links: |
ANKRD9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANKRD9 - ankyrin repeat domain 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152326.2 | 1428 | Missense Mutation | ATG,CTG | M,L 232 | NP_689539.1 | |
XM_005267310.2 | 1428 | Missense Mutation | ATG,CTG | M,L 232 | XP_005267367.1 | |
XM_005267311.3 | 1428 | Missense Mutation | ATG,CTG | M,L 232 | XP_005267368.1 |
TECPR2 - tectonin beta-propeller repeat containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |