Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGCAAGGCATATTTACCTGTGGC[A/C]TGCTGTGTGCCCGGCACTGTTCTGA
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 182350 MIM: 600283 | ||||||||||||||||||||||||||
Literature Links: |
ATP1A3 PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
ATP1A3 - ATPase Na+/K+ transporting subunit alpha 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GRIK5 - glutamate ionotropic receptor kainate type subunit 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301030.1 | Intron | NP_001287959.1 | ||||
NM_002088.4 | Intron | NP_002079.3 | ||||
XM_005258821.3 | Intron | XP_005258878.1 | ||||
XM_011526862.2 | Intron | XP_011525164.1 | ||||
XM_011526863.2 | Intron | XP_011525165.1 | ||||
XM_011526865.1 | Intron | XP_011525167.1 | ||||
XM_011526867.1 | Intron | XP_011525169.1 | ||||
XM_011526868.1 | Intron | XP_011525170.1 | ||||
XM_011526869.2 | Intron | XP_011525171.1 | ||||
XM_011526870.2 | Intron | XP_011525172.1 | ||||
XM_011526871.2 | Intron | XP_011525173.1 | ||||
XM_017026713.1 | Intron | XP_016882202.1 |
Set Membership: |
HapMap |