Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACACTGAGTAGACTTGTCATTAT[C/T]GGAAGCTGATCCAAGTGCTAAAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612636 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UNC80 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
UNC80 - unc-80 homolog, NALCN activator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032504.1 | Intron | NP_115893.1 | ||||
NM_182587.3 | Intron | NP_872393.3 | ||||
XM_005246476.1 | Intron | XP_005246533.1 | ||||
XM_011511004.1 | Intron | XP_011509306.1 | ||||
XM_011511005.1 | Intron | XP_011509307.1 | ||||
XM_011511006.1 | Intron | XP_011509308.1 | ||||
XM_011511007.1 | Intron | XP_011509309.1 | ||||
XM_011511008.1 | Intron | XP_011509310.1 | ||||
XM_011511010.2 | Intron | XP_011509312.1 | ||||
XM_011511012.1 | Intron | XP_011509314.1 | ||||
XM_017003884.1 | Intron | XP_016859373.1 | ||||
XM_017003885.1 | Intron | XP_016859374.1 | ||||
XM_017003886.1 | Intron | XP_016859375.1 | ||||
XM_017003887.1 | Intron | XP_016859376.1 | ||||
XM_017003888.1 | Intron | XP_016859377.1 | ||||
XM_017003889.1 | Intron | XP_016859378.1 | ||||
XM_017003890.1 | Intron | XP_016859379.1 | ||||
XM_017003891.1 | Intron | XP_016859380.1 | ||||
XM_017003892.1 | Intron | XP_016859381.1 | ||||
XM_017003893.1 | Intron | XP_016859382.1 | ||||
XM_017003894.1 | Intron | XP_016859383.1 |