Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAGGTTTTCCAGGTCCTGGATAT[A/G]GCAGCGTAAGATGGGAAATCACAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605464 MIM: 611741 MIM: 123831 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCB8 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ABCB8 - ATP binding cassette subfamily B member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282291.1 | Intron | NP_001269220.1 | ||||
NM_001282292.1 | Intron | NP_001269221.1 | ||||
NM_001282293.1 | Intron | NP_001269222.1 | ||||
NM_007188.4 | Intron | NP_009119.2 |
ASIC3 - acid sensing ion channel subunit 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004769.3 | Intron | NP_004760.1 | ||||
NM_020321.3 | Intron | NP_064717.1 | ||||
NM_020322.3 | Intron | NP_064718.1 |
CDK5 - cyclin dependent kinase 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |