Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTATTTGTATTGTTATTGTATTG[G/T]TTCTCATTTTTTTCTATTTTTTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 400033 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBL1Y PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TBL1Y - transducin (beta)-like 1, Y-linked | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033284.1 | Intron | NP_150600.1 | ||||
NM_134258.1 | Intron | NP_599020.1 | ||||
NM_134259.1 | Intron | NP_599021.1 | ||||
XM_005262572.3 | Intron | XP_005262629.1 | ||||
XM_017030086.1 | Intron | XP_016885575.1 | ||||
XM_017030087.1 | Intron | XP_016885576.1 |