Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAGTCCAGGGGACGGCTAGAGGT[A/G]AGGTCAGCCGCAAGACTGGCGTCAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCSER2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCSER2 - coiled-coil serine rich protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284240.1 | Intron | NP_001271169.1 | ||||
NM_001284241.1 | Intron | NP_001271170.1 | ||||
NM_001284242.1 | Intron | NP_001271171.1 | ||||
NM_001284243.1 | Intron | NP_001271172.1 | ||||
NM_018999.3 | Intron | NP_061872.2 | ||||
XM_005269905.3 | Intron | XP_005269962.1 | ||||
XM_005269906.3 | Intron | XP_005269963.1 | ||||
XM_005269908.3 | Intron | XP_005269965.1 | ||||
XM_006717894.1 | Intron | XP_006717957.1 | ||||
XM_006717895.1 | Intron | XP_006717958.1 | ||||
XM_011539870.2 | Intron | XP_011538172.1 | ||||
XM_011539871.2 | Intron | XP_011538173.1 | ||||
XM_011539872.2 | Intron | XP_011538174.1 | ||||
XM_011539873.1 | Intron | XP_011538175.1 | ||||
XM_017016340.1 | Intron | XP_016871829.1 | ||||
XM_017016341.1 | Intron | XP_016871830.1 | ||||
XM_017016342.1 | Intron | XP_016871831.1 | ||||
XM_017016343.1 | Intron | XP_016871832.1 |