Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCAKD PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DCAKD - dephospho-CoA kinase domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128631.2 | 2022 | UTR 3 | NP_001122103.1 | |||
NM_001288654.2 | 2022 | UTR 3 | NP_001275583.1 | |||
NM_001288655.1 | 2022 | UTR 3 | NP_001275584.1 | |||
NM_001321326.1 | 2022 | UTR 3 | NP_001308255.1 | |||
NM_024819.6 | 2022 | UTR 3 | NP_079095.3 | |||
XM_005257688.2 | 2022 | Intron | XP_005257745.1 | |||
XM_011525262.2 | 2022 | UTR 3 | XP_011523564.1 | |||
XM_017025102.1 | 2022 | Intron | XP_016880591.1 | |||
XM_017025103.1 | 2022 | Intron | XP_016880592.1 | |||
XM_017025104.1 | 2022 | Intron | XP_016880593.1 | |||
XM_017025105.1 | 2022 | UTR 3 | XP_016880594.1 | |||
XM_017025106.1 | 2022 | UTR 3 | XP_016880595.1 |
Set Membership: |
HapMap |