Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612027 MIM: 139139 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRASP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRASP - general receptor for phosphoinositides 1 associated scaffold protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NR4A1 - nuclear receptor subfamily 4 group A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202233.1 | Intron | NP_001189162.1 | ||||
NM_001202234.1 | Intron | NP_001189163.1 | ||||
NM_002135.4 | Intron | NP_002126.2 | ||||
NM_173157.2 | Intron | NP_775180.1 | ||||
XM_005268822.3 | Intron | XP_005268879.1 | ||||
XM_005268824.3 | Intron | XP_005268881.1 | ||||
XM_006719363.1 | Intron | XP_006719426.1 | ||||
XM_006719364.3 | Intron | XP_006719427.1 | ||||
XM_017019247.1 | Intron | XP_016874736.1 | ||||
XM_017019248.1 | Intron | XP_016874737.1 | ||||
XM_017019249.1 | Intron | XP_016874738.1 |