Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGGGGAGGGCCCCTGGCCAGACA[T/C]GGACGGCGCCATGGTTGAGCAGAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615054 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ASB10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ASB10 - ankyrin repeat and SOCS box containing 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142459.1 | 1340 | Missense Mutation | CAT,CGT | H,R 360 | NP_001135931.2 | |
NM_001142460.1 | 1340 | Missense Mutation | CAT,CGT | H,R 360 | NP_001135932.2 | |
NM_080871.3 | 1340 | Missense Mutation | CAT,CGT | H,R 345 | NP_543147.2 |
IQCA1L - IQ motif containing with AAA domain 1 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |