Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602673 MIM: 604434 MIM: 605504 | ||||||||||||||||||||||||||||||||
Literature Links: |
KLK10 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
KLK10 - kallikrein related peptidase 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077500.1 | 1399 | UTR 3 | NP_001070968.1 | |||
NM_002776.4 | 1399 | UTR 3 | NP_002767.2 | |||
NM_145888.2 | 1399 | UTR 3 | NP_665895.1 | |||
XM_005259061.3 | 1399 | UTR 3 | XP_005259118.1 | |||
XM_005259062.3 | 1399 | UTR 3 | XP_005259119.1 | |||
XM_006723287.3 | 1399 | UTR 3 | XP_006723350.1 | |||
XM_006723289.3 | 1399 | UTR 3 | XP_006723352.1 | |||
XM_017026993.1 | 1399 | UTR 3 | XP_016882482.1 |
KLK11 - kallikrein related peptidase 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KLK9 - kallikrein related peptidase 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |