Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTCTTCTCATCTTTCCCCTGCT[C/T]ATCTGAGTCAGAGCTGGAACTGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C14orf105 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C14orf105 - chromosome 14 open reading frame 105 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001283056.1 | 1067 | Missense Mutation | AAG,GAG | K,E 307 | NP_001269985.1 | |
NM_001283057.1 | 1067 | Missense Mutation | AAG,GAG | K,E 266 | NP_001269986.1 | |
NM_001283058.1 | 1067 | Missense Mutation | AAG,GAG | K,E 178 | NP_001269987.1 | |
NM_001283059.1 | 1067 | Missense Mutation | AAG,GAG | K,E 178 | NP_001269988.1 | |
NM_001283060.1 | 1067 | Intron | NP_001269989.1 | |||
NM_018168.3 | 1067 | Missense Mutation | AAG,GAG | K,E 267 | NP_060638.2 | |
XM_005267806.1 | 1067 | Intron | XP_005267863.1 | |||
XM_005267810.3 | 1067 | Missense Mutation | AAG,GAG | K,E 188 | XP_005267867.1 | |
XM_005267811.3 | 1067 | Missense Mutation | AAG,GAG | K,E 179 | XP_005267868.1 | |
XM_005267813.1 | 1067 | Missense Mutation | AAG,GAG | K,E 141 | XP_005267870.1 | |
XM_005267814.1 | 1067 | Missense Mutation | AAG,GAG | K,E 140 | XP_005267871.1 | |
XM_006720188.2 | 1067 | Intron | XP_006720251.1 | |||
XM_006720189.2 | 1067 | Missense Mutation | AAG,GAG | K,E 189 | XP_006720252.1 |