Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGTCCGCCCCTGCGGCGCGTCGC[A/C]GTGTGGCCGGTGTTGCGAGAGCGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602360 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GATM PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GATM - glycine amidinotransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321015.1 | 442 | Intron | NP_001307944.1 | |||
NM_001482.2 | 442 | Intron | NP_001473.1 |
SPATA5L1 - spermatogenesis associated 5 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323640.1 | 442 | Silent Mutation | GCA,GCC | A,A 111 | NP_001310569.1 | |
NM_024063.2 | 442 | Silent Mutation | GCA,GCC | A,A 111 | NP_076968.2 |