Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCATCCTCAGAGCTGGCTTCACA[A/G]GCAGTGTGGAAGAGCTGCATGAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610393 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GON4L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GON4L - gon-4 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282856.1 | 6757 | Silent Mutation | GCC,GCT | A,A 2207 | NP_001269785.1 | |
NM_001282858.1 | 6757 | Silent Mutation | GCC,GCT | A,A 2206 | NP_001269787.1 | |
NM_001282860.1 | 6757 | Silent Mutation | GCC,GCT | A,A 2207 | NP_001269789.1 | |
NM_001282861.1 | 6757 | Intron | NP_001269790.1 | |||
NM_032292.5 | 6757 | Intron | NP_115668.4 | |||
XM_005245284.3 | 6757 | Silent Mutation | GCC,GCT | A,A 2034 | XP_005245341.1 | |
XM_005245286.3 | 6757 | Silent Mutation | GCC,GCT | A,A 1402 | XP_005245343.1 | |
XM_006711393.3 | 6757 | Silent Mutation | GCC,GCT | A,A 2206 | XP_006711456.1 | |
XM_006711394.3 | 6757 | Silent Mutation | GCC,GCT | A,A 2206 | XP_006711457.1 | |
XM_011509658.2 | 6757 | Silent Mutation | GCC,GCT | A,A 2177 | XP_011507960.1 | |
XM_011509659.2 | 6757 | Silent Mutation | GCC,GCT | A,A 2034 | XP_011507961.1 |
LOC100505728 - uncharacterized LOC100505728 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MSTO2P - misato family member 2, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |