Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGTGTTTGGGGTGCTCCGCTCCT[C/T]TGGGGTCTTCTTCGTGGAGTTTGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608992 MIM: 615765 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCL6B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCL6B - B-cell CLL/lymphoma 6B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC16A11 - solute carrier family 16 member 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC16A13 - solute carrier family 16 member 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_201566.2 | 409 | Missense Mutation | TCT,TTT | S,F 34 | NP_963860.1 |