Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118820 MIM: 139240 MIM: 612756 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSH2 - chorionic somatomammotropin hormone 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020991.3 | Intron | NP_066271.1 | ||||
NM_022644.3 | Intron | NP_072170.1 | ||||
NM_022645.2 | Intron | NP_072171.1 |
GH2 - growth hormone 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TCAM1P - testicular cell adhesion molecule 1, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |