Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGAACCCCAAAAGCAGGGAGGT[A/G]CAGAGAGCCATGAAGCTCCTGGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602565 MIM: 608529 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCL25 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCL25 - C-C motif chemokine ligand 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201359.1 | 360 | Silent Mutation | GTA,GTG | V,V 83 | NP_001188288.1 | |
NM_005624.3 | 360 | Silent Mutation | GTA,GTG | V,V 83 | NP_005615.2 | |
XM_011528177.2 | 360 | Silent Mutation | GTA,GTG | V,V 83 | XP_011526479.1 | |
XM_017027119.1 | 360 | Silent Mutation | GTA,GTG | V,V 143 | XP_016882608.1 | |
XM_017027120.1 | 360 | Silent Mutation | GTA,GTG | V,V 83 | XP_016882609.1 | |
XM_017027121.1 | 360 | Silent Mutation | GTA,GTG | V,V 83 | XP_016882610.1 | |
XM_017027122.1 | 360 | Silent Mutation | GTA,GTG | V,V 83 | XP_016882611.1 |
FBN3 - fibrillin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |