Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGCAGAACGCAGTTTGCAACATA[G/T]CAGGTAGGCCCGAACCTGGCAGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612012 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZFYVE26 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZFYVE26 - zinc finger FYVE-type containing 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015346.3 | 5139 | Missense Mutation | ATA,CTA | I,L 2478 | NP_056161.2 | |
XM_011536609.2 | 5139 | Intron | XP_011534911.1 | |||
XM_017021124.1 | 5139 | Missense Mutation | ATA,CTA | I,L 2484 | XP_016876613.1 | |
XM_017021125.1 | 5139 | Intron | XP_016876614.1 | |||
XM_017021126.1 | 5139 | Missense Mutation | ATA,CTA | I,L 1981 | XP_016876615.1 | |
XM_017021127.1 | 5139 | Missense Mutation | ATA,CTA | I,L 1709 | XP_016876616.1 | |
XM_017021128.1 | 5139 | Missense Mutation | ATA,CTA | I,L 1678 | XP_016876617.1 |